Thermo Scientific ND-3300 Fluorospectrometer 83060-50
Representative image only.
Thermo Scientific Mfr # ND-3300 – Item # EW-83060-50
Saves time and space while utilizing as little as 1 µL of sample
  • Pipette your sample directly onto the optical surface — no need for separate cuvettes
  • Obtain readings in as little as 5 seconds!
  • Small footprint works gerat for even the most space-limited lab
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Source LampLED
  • Min Wavelength (nm)400
  • Max Wavelength (nm)750
  • Wavelength Accuracy±1.0 nm
  • Detector2048-element linear silicon CCD array
  • Scan Time<10 seconds
  • DescriptionFluorospectrometer 83060-50
  • Warranty1 year
1 Year Warranty
Key Features
  • Pipette your sample directly onto the optical surface — no need for separate cuvettes
  • Obtain readings in as little as 5 seconds!
  • Small footprint works gerat for even the most space-limited lab
  • 1 year warranty
More About this Item
The Thermo Scientific NanoDrop fluorospectrometer provides fluorescent analysis of 1 µL samples from 400 to 750 nm. Unit excites the sample from one of three LED sources; UV, blue, or white. The direct coupling of the sample to the optics and the proprietary signal processing for white LED applications enable measurements across a wide range of wavelengths — no need for filter changes.
The small sample size allows you to minimize loss, making units ideal for precious samples. Eliminating the need for cuvettes helps reduce error in results, giving you confidence that the results obtained are accurate.
The fluorospectrometer base measures only 5-1/2” x 7-7/8” (14.0 x 20.0 cm), taking up minimal lab space.
Includes software; simply plug the instrument into your computer and you’re ready to go!
Available for sale and use in the United States only.
Product may contain trace amounts of mercury; cautions should be used in breakage, cleanup, shipping and/or disposal.