Thermo Scientific ND-NDL-US-CAN Spectrophotometer 120V

Representative image only.
Thermo Scientific Mfr # ND-NDL-US-CAN – Item # EW-83060-01
Boasts a tiny footprint yet is powerful enough for routine measurements in life science applications
  • Stand-alone unit does not require a PC — small enough to fit in a drawer!
  • Small sample size requirements (1 to 2 µL) preserves your important samples
  • Superfast measurements saves time
  • Measure DNA, RNA and purified protein measurements in addition to nucleic acid samples

$5,500.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Source LampLED
  • Min Wavelength (nm)260
  • Max Wavelength (nm)280
  • Min Photometric - Absorbance (A)0
  • Max Photometric - Absorbance (A)30
  • DetectorSilicon photodiode
  • Concentration0-1500 ng/µL (dsDNA); 0-45 mg/mL (BSA)
  • Materials of Construction304 Stainless Steel, quartz fiber
  • Width (cm)11.5
  • Depth (cm)16
  • Width (in)4 1/2
  • Depth (in)6 19/64
  • Power (VDC)6
  • DescriptionSpectrophotometer 120V

Key Features

  • Stand-alone unit does not require a PC — small enough to fit in a drawer!
  • Small sample size requirements (1 to 2 µL) preserves your important samples
  • Superfast measurements saves time
  • Measure DNA, RNA and purified protein measurements in addition to nucleic acid samples

More About this Item

The smallest UV spectrophotometer in the Nanodrop family, the Nanodrop Lite was designed specifically for microvolume measurements of purified nucleic acids and proteins. It measures nucleic acid concentrations at 260 nm, purity ratio at 260/280nm, and purified protein concentrations at 280 nm. No plates or cuvettes are required, saving you time and money.

Simply pipette your sample using direct measurement or place sample on the stainless steel pedestal. The unique sample-retention system uses surface tension and requires only 1 to 2 µL of sample without the need for dilution — saves time while reducing waste. Highly polished quartz and stainless steel surfaces are resistant to sample adherence, making the use of dry lab wipes very effective in removing the sample in preparation for your next measurement.

This stand-alone, local control unit does not require a PC and comes loaded with four pre-configured applications for fast out-of-box setup. You can easily store the last 500 measurements taken or export your data to a PC via the USB. Print measurement information on cryogenic labels using printer 83060-51 (sold separately below). For added convenience, Nanodrop Lite is also available with a printer or PC.

Available for sale in the United States only

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat