Thermo Scientific ND3300LAPTOP Fluorospectrometer with Laptop

Representative image only.
Thermo Scientific Mfr # ND3300LAPTOP – Item # EW-83061-56
Saves time and space while utilizing as little as 1 µL of sample
  • Pipette your sample directly onto the optical surface — no need for separate cuvettes
  • Obtain readings in as little as 5 seconds!
  • Small footprint works gerat for even the most space-limited lab

PRODUCT OPTIONS

$15,219.51USD / Each
$15,600.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Source LampLED
  • Min Wavelength (nm)260
  • Max Wavelength (nm)280
  • Min Photometric - Absorbance (A)0
  • Max Photometric - Absorbance (A)30
  • DetectorSilicon photodiode
  • Concentration0-1500 ng/µL (dsDNA); 0-45 mg/mL (BSA)
  • Materials of Construction304 Stainless Steel, quartz fiber
  • Width (cm)11.5
  • Depth (cm)16
  • Width (in)4 1/2
  • Depth (in)6 19/64
  • Power (VDC)6
  • DescriptionFluorospectrometer with Laptop

Key Features

  • Pipette your sample directly onto the optical surface — no need for separate cuvettes
  • Obtain readings in as little as 5 seconds!
  • Small footprint works gerat for even the most space-limited lab

More About this Item

The Thermo Scientific NanoDrop fluorospectrometer provides fluorescent analysis of 1 µL samples from 400 to 750 nm. Unit excites the sample from one of three LED sources; UV, blue, or white. The direct coupling of the sample to the optics and the proprietary signal processing for white LED applications enable measurements across a wide range of wavelengths — no need for filter changes.
The small sample size allows you to minimize loss, making units ideal for precious samples. Eliminating the need for cuvettes helps reduce error in results, giving you confidence that the results obtained are accurate.
The fluorospectrometer base measures only 5-1/2” x 7-7/8” (14.0 x 20.0 cm), taking up minimal lab space.

Includes software and computer—you’re ready to go!

Available for sale in the US only.

Product may contain trace amounts of mercury; cautions should be used in breakage, cleanup, shipping and/or disposal.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat