Thermo Scientific Nunc 136196 Nunclon cell culture flask, 25 cm2, angled neck, filter cap
Representative image only.
Thermo Scientific Nunc Mfr # 136196 – Item # EW-01932-41
Nunclon® Delta surface treatment provides optimal cell attachment and growth
  • Virgin polystyrene provides excellent optical quality
  • Sterilized by gamma radiation
  • Large-bore necks—easy pipette and cell scraper access
PRODUCT OPTIONS
$565.00USD / Case of 160
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolystyrene
  • Cap TypeFilter
  • Cap MaterialHDPE
  • Neck TypeAngled
  • Total Volume (mL)50
  • Culture Area (cm²)25
  • DescriptionNunclon cell culture flask, 25 cm2, angled neck, filter cap
Key Features
  • Virgin polystyrene provides excellent optical quality
  • Sterilized by gamma radiation
  • Large-bore necks—easy pipette and cell scraper access
Call Us At1-847-549-7600
Live chat