Thermo Scientific Nunc Cell Culture Multidishes, Treated, Polystyrene, Sterile, 6-Well, 3 mL; 85/Cs
Representative image only.
Thermo Scientific Nunc Mfr # 140685 – Item # EW-01930-21
Nunclon certified surface ensures optimum cell attachment and growth—use for scale up, cloning, viral titration, and fusion
  • Sterilized by gamma irradiation and certified to be non-pyrogenic
  • Tested for cell growth and plating efficiency to ensure consistent biological performance
  • Flat well bottom and stackable
  • Raised well rims lower the risk of cross contamination
  • Alphanumerical codes for well identification
PRODUCT OPTIONS
$386.00USD / Case of 85
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells6
  • Max Working Well Volume (mL)3
  • Well Diameter (mm)35
  • Well ShapeRound
  • Culture Area (cm²)9.6
  • Surface TreatmentNunclon™ Delta
  • Lid (Included)Yes
  • SterileYes
  • DescriptionNunclon 6-Well PS MultiDish w/Lid, Round; 85/Cs
Sterile
Key Features
  • Sterilized by gamma irradiation and certified to be non-pyrogenic
  • Tested for cell growth and plating efficiency to ensure consistent biological performance
  • Flat well bottom and stackable
  • Raised well rims lower the risk of cross contamination
  • Alphanumerical codes for well identification
More About this Item
Round well styles feature vented lids for gas exchange and recessed areas around wells that act as a water reservoir to reduce evaporation from wells. Dishes with 6, 12, 24, and 48-well formats include lids with condensation rings to minimize evaporation. The 12 and 48-well dishes have a standard 96-well footprint. Rectangular well styles offer a larger culture area. They also accept standard glass cover slips: 24 x 60mm for 4-well plate; and 22 x 22mm for 8 well plate. Radiation sterilized.