Thermo Scientific Nunc Cell Culture Petri Dishes, Nunclon™ Delta Treated, Sterile, Non-Gridded, 60 x 15 mm; 400/Cs
Representative image only.
Thermo Scientific Nunc Mfr # 150288 – Item # EW-01930-32
Nunclon™ Delta surface treatment promotes maximum adhesion for a broad range of cell types
  • Top and bottom stacking rings for easier handling and more stable stacking
  • Sterile and non-pyrogenic
  • Constructed of durable polystyrene
  • Lid features vented for air flow
PRODUCT OPTIONS
$478.00USD / Case of 400
SPECS & DESCRIPTION
Specifications & Description
  • Max Working Well Volume (mL)5
  • Well Diameter (mm)60
  • Well ShapeRound
  • Culture Area (cm²)21.5
  • Surface TreatmentNunclon™ Delta
  • Lid (Included)Yes, Vented
  • SterileYes
  • DescriptionCell Culture Petri Dishes, Nunclon™ Delta Treated, Sterile, Non-Gridded, 60 x 15 mm; 400/Cs
Key Features
  • Top and bottom stacking rings for easier handling and more stable stacking
  • Sterile and non-pyrogenic
  • Constructed of durable polystyrene
  • Lid features vented for air flow
More About this Item
Perform a variety of applications in dishes with the Nunclon™ Delta surface treatment that promotes maximum adhesion for a broad range of cell types. Petri dishes are vented for air flow. Gridded model options feature a grid bottom specially designed for cloning applications and determining plating efficiencies. All dishes are sterilized by gamma irradiation.