Thermo Scientific Nunc EasyFlask™ Cell Culture Flask, Polystyrene, Sterile, Non-Treated, Angled Neck, 175 cm2; 30/Cs
Representative image only.
Thermo Scientific Nunc Mfr # 159926 – Item # EW-01928-74
Non-treated surface offers low cell attachment
  • Certified nonpyrogenic and sterile
  • White caps for easy identification
  • The filter cap ensures constant airflow while minimizing chances of contamination
  • EasyFlask™ allows for easy handling and pipetting
PRODUCT OPTIONS
$322.00USD / Case of 30
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolystyrene
  • Cap TypeFilter
  • Cap MaterialHDPE
  • Neck TypeAngled
  • Working Volume (mL)55
  • Total Volume (mL)645
  • Culture Area (cm²)175
  • DescriptionEasyFlask™ Cell Culture Flask, Angled Neck, 175 cm2; 30/Cs
Nonpyrogenic
Key Features
  • Certified nonpyrogenic and sterile
  • White caps for easy identification
  • The filter cap ensures constant airflow while minimizing chances of contamination
  • EasyFlask™ allows for easy handling and pipetting
More About this Item
These sterile culture flasks are used for unique cell growth applications that do not require a Nunclon™ Delta surface treatment. Untreated surface offers low attachment. Clear, uniform design provides excellent optical quality, making it a suitable choice for microscopy. Flasks have a white cap for easy identification. All models are certified nonpyrogenic and sterile.
TripleFlask™ model 01928-42 offers a unique three-parallel growth surface that provides a total culture area of 500 cm2 - ideal for scale-up.
EasyFlask™ models 01928-70, -72, and -74 feature an ergonomic design requiring only a 1/3 turn to open and close the cap.