Thermo Scientific Nunc Edge™ Microplates with Lid, 96-Well, Polystyrene, Sterile, Nunclon™ Treated; 50/Cs

Representative image only.
Thermo Scientific Nunc Mfr # 167425 – Item # EW-01930-64
Minimize plate evaporation and the risks associated with edge effect
  • Excellent optical performance
  • Reduced edge effects
  • Use 38% more of your plate – cells are still viable
  • Nunclon™ Delta cell culture surface treatment of the wells promotes cell attachment and growth

PRODUCT OPTIONS

$500.00USD / Case of 50
Often Bought With…

SPECS & DESCRIPTION

Specifications & Description

  • Number of Wells96
  • Well Volume (µL)400
  • Well Bottom Diameter (mm)6.7
  • Well Top Diameter (mm)7.1
  • Well ShapeFlat
  • Surface TreatmentNunclon™ Delta
  • Lid (Included)Yes
  • SterileYes
  • Plate ColorClear
  • MaterialPolystyrene
  • Width (mm)86
  • Length (mm)128
  • DescriptionMicroplates with Lid, 96-Well, Polystyrene, Sterile, Nunclon™ Treated; 50/Cs

Key Features

  • Excellent optical performance
  • Reduced edge effects
  • Use 38% more of your plate – cells are still viable
  • Nunclon™ Delta cell culture surface treatment of the wells promotes cell attachment and growth

More About this Item

The Edge™ 2.0 96-well plates provide an efficient and economical solution for preventing evaporation and in turn the risks associated with edge effect. The surrounding built-in moat is divided into 4 sectional reservoirs, each can be filled with 1.7 to 2.0mL of sterile water, buffer, or media. When filled with fluid, the surrounding moat acts as an evaporation buffer during extended incubation, yielding consistent, viable cells across the entire 96-well plate that in turn enhances productivity for cell-based assays. Enabling the use of all 96 wells allows researchers to plan experiments more efficiently, resulting in potentially less cost and related product waste. Choose from Nunclon™ treated or non-treated surfaces.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat