Thermo Scientific Nunc 263339 Nonsterile lids for 96-well plates 01929-32, -33, -34, and -38
Representative image only.
Thermo Scientific Nunc Mfr # 263339 – Item # EW-01929-10
Secure samples within your 96-well microplates
  • Protect samples from contamination and evaporation during assay processing, incubation or storage
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionNonsterile lids for 96-well plates 01929-32, -33, -34, and -38
Key Features
  • Protect samples from contamination and evaporation during assay processing, incubation or storage
More About this Item
Choose from a range of options to prevent contamination and evaporation of well contents and safely store samples within your 96-well plates.
Lids have notices for quick orientation and have individual condensation rings for enhanced sample security.
Sealing tap is an adhesive seal that effectively seals all microplate formats. Pressure sensitive adhesive is not tacky to touch but forms a strong bond to the plate upon application of pressure.
Cap Mats create a tight seal but are easy to peel off without aerosols—reducing the risk of well-to-well contamination. Caps are pierceable by hypodermic needles without compromising seal.