Thermo Scientific Nunc 339650 Conical Centrifuge Tubes 500/CS, 15 mL
Representative image only.
Thermo Scientific Nunc Mfr # 339650 – Item # EW-22999-80
Developed with your research needs in mind!
  • Reusable, collapsible and recyclable plastic rack is environmentally friendly
  • Large writing area provides ample space for labeling your samples
  • High RCF rating enables greater range of use and applications
PRODUCT OPTIONS
$302.00USD / Case of 500
SPECS & DESCRIPTION
Specifications & Description
  • Max RCF x g10500
  • SterileYes
  • Tube Capacity (mL)15
  • Tube MaterialPolypropylene
  • Tube ColorClear
  • Tube Bottom ShapeConical
  • Cap IncludedYes
  • Cap MaterialHDPE
  • GraduationsYes
  • DescriptionConical Centrifuge Tubes 500/CS, 15 mL
Key Features
  • Reusable, collapsible and recyclable plastic rack is environmentally friendly
  • Large writing area provides ample space for labeling your samples
  • High RCF rating enables greater range of use and applications
More About this Item
These conical-bottom centrifuge tubes were developed for function and flexibility. The tubes provide a large writing space to label your samples. Polypropylene material is durable and can handle high centrifugation speeds, allowing for greater range of use in the lab. The plastic rack is collapsible and will break down when not in use, saving valuable lab space. The rack is manufactured with recyclable materials, lessening environmental impact and reducing waste in the lab.