Thermo Scientific Nunc 377224 CryoTubes, Internal Thread/Starfoot/1.0 mL; 50/Pk

Representative image only.
Thermo Scientific Nunc Mfr # 377224 – Item # EW-03757-60
  • Polyethylene closure with silicone gasket provides a secure seal.

PRODUCT OPTIONS

$62.50USD / Pkg of 50

SPECS & DESCRIPTION

Specifications & Description

  • Volume (mL)1
  • Tube MaterialPolypropylene
  • Tube Bottom ShapeConical
  • Cap TypeScrew-Thread
  • ThreadInternal
  • SterileYes
  • Height (in)1 21/32
  • Outside Diameter (in)31/64
  • Height (mm)42
  • Outside Diameter (mm)12.5
  • DescriptionCryoTubes, Internal Thread/Starfoot/1.0 mL; 50/Pk

Key Features

  • Polyethylene closure with silicone gasket provides a secure seal.

More About this Item

Presterilized by cobalt-60 radiation, these one-piece molded polypropylene vials withstand temperatures from -196 to 135°C. Starfooted vials lock into CryoTube® vial holder 03755-71 for one-handed operation. Optional colored cap inserts simplify sample identification.
Note: Do not autoclave or immerse vials in liquid nitrogen. For storage under extreme conditions such as liquid nitrogen, use CryoFlex™ tubing 03755-64.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat