Thermo Scientific Nunc Cell Culture Multidishes, Treated, Polystyrene, Sterile, 48-Well, 0.5 mL; 75/Cs
Representative image only.
Thermo Scientific Nunc Mfr # 150687 – Item # EW-01930-24
Nunclon certified surface ensures optimum cell attachment and growth—use for scale up, cloning, viral titration, and fusion
  • Sterilized by gamma irradiation and certified to be non-pyrogenic
  • Tested for cell growth and plating efficiency to ensure consistent biological performance
  • Flat well bottom and stackable
  • Raised well rims lower the risk of cross contamination
  • Alphanumerical codes for well identification
  • 1 year warranty
PRODUCT OPTIONS
$491.00USD / Case of 75
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells48
  • Max Working Well Volume (mL)0.5
  • Well Diameter (mm)12
  • Well ShapeRound
  • Culture Area (cm²)1.1
  • Surface TreatmentNunclon™ Delta
  • Lid (Included)Yes
  • SterileYes
  • DescriptionNunclon 48-Well PS MultiDish w/Lid, Round; 75/Cs
  • Warranty1 year
1 Year WarrantySterile
Key Features
  • Sterilized by gamma irradiation and certified to be non-pyrogenic
  • Tested for cell growth and plating efficiency to ensure consistent biological performance
  • Flat well bottom and stackable
  • Raised well rims lower the risk of cross contamination
  • Alphanumerical codes for well identification
  • 1 year warranty
More About this Item
Round well styles feature vented lids for gas exchange and recessed areas around wells that act as a water reservoir to reduce evaporation from wells. Dishes with 6, 12, 24, and 48-well formats include lids with condensation rings to minimize evaporation. The 12 and 48-well dishes have a standard 96-well footprint. Rectangular well styles offer a larger culture area. They also accept standard glass cover slips: 24 x 60mm for 4-well plate; and 22 x 22mm for 8 well plate. Radiation sterilized.