This item is now discontinued. We recommend the following as suggested replacements: EW-12152-00
Thermo Scientific Orbital motion heated bath; 120V
Representative image only.
Thermo Scientific Mfr # 3540 – Item # EW-01290-20
Energy saving—three position heating switch Separate heating and shaking controls for versatility
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Bath TypeShaking Water Bath
  • Reservoir Capacity (L)18
  • Max Temperature (° C)65
  • Min Temperature (° C)Ambient
  • Temperature Stability (° C)±0.5
  • High-Temperature CutoffYes
  • Heater Wattage (watts)1150
  • Temperature SensorHydraulic
  • Wetted Materials304 SS
  • Power (VAC)115
  • DescriptionOrbital motion heated bath; 120V
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
This orbital-motion heated bath features a built-in shaker timer that can be set in 1-minute increments up to 60 minutes, or set for continuous operation. A 1/15-hp induction motor produces a 1/2" circular motion; 40 to 400 rpm. Analog tachometer indicates speed.
Water bath chamber is made of seamless, drawn stainless steel and is insulated with a 2" thick blanket of fiberglass. Perforated stainless steel baffles minimize splashing. Outer housing is polyethylene. Bath includes a 6-ft siphon hose. Order platforms (sold separately below) to hold your glassware.