Thermo Scientific 1115000 4-star benchtop pH/ISE meter, 115/220V

Representative image only.
Thermo Scientific Mfr # 1115000 – Item # EW-58823-04
Redesigned meters feature auto-sampler compatibility, improved accuracy, additional security, and datalog capabilities, additional temperature calibrations, and many more new functions!
  • Improved 4-Star meters are ideal for labs with strict calibration, measurement, and datalogging requirements
  • Available 21 CFR Part 11 software for FDA regulatory compliance (order separately to the right)
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeBenchtop
  • Display typeLCD
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.002 pH
  • Buffer RecognitionDIN, NIST, USA, and custom
  • CalibrationUp to 5 points
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution0.1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min ISE0 to 19,990 ppm
  • Temperature compensationAutomatic or manual
  • WaterproofYes
  • Width (cm)16.8275
  • Height (cm)9.525
  • Width (in)6 5/8
  • Height (in)3 3/4
  • Description4-star benchtop pH/ISE meter, 115/220V
  • Warranty3 year
3 Year WarrantyConformite EuropeenneCanadian Standards AssociationIP54 RatedISO9001:2000 Certified SupplierUL Listed.

Key Features

  • Improved 4-Star meters are ideal for labs with strict calibration, measurement, and datalogging requirements
  • Available 21 CFR Part 11 software for FDA regulatory compliance (order separately to the right)
  • 3 year warranty

More About this Item

All Orion Star Series meters include a convenient Quick-Start guide to help you get started and provide support while in use. Smart Stability™ and Smart Averaging™ ensure accurate results. GLP-related features including datalogging with time/date stamp and electrode verification. Meters feature automatic or manual pH calibration, auto-buffer recognition, advanced self-tests and selfdiagnostics,
and RS-232 or optional USB connectivity.

These 4-Star benchtop meters now include an improved stability algorithm to increase calibration and measurement accuracy. Meters can now be paired with autosamplers for increased sample throughput. Increased memory allows you to log more data points and even turn off the data log if desired. Improved security is offered by new individual methods passwords. Temperature accuracy can be ensured with the ATC temperature calibration feature.

Meters include: Quick-Start guide and universal power supply.

Meter kits add: ROSS Ultra® pH electrode, epoxy ATC probe, buffers (one 475-mL bottle each of pH 4.01, 7.00, and 10.00), pH electrode storage solution, and benchtop electrode stand.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat