Thermo Scientific 1215000 Optional pH/ISE Meter

Representative image only.
Thermo Scientific Mfr # 1215000 – Item # EW-58822-04
Enhanced capabilities for improved accuracy, additional security, increased datalog capabilities, temperature calibrations, and more!
  • Improved stability algorithm increases calibration and measurement accuracy
  • Ensure temperature accuracy by performing the ATC temperature calibration
  • Increased memory allows you to log more data points or turn off the data log if desired
  • Print or view your last 10 calibrations
  • New individual method passwords provide better security
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypePortable
  • Display typeLCD
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.002 pH
  • Buffer RecognitionDIN, NIST, USA, and custom
  • CalibrationUp to 5 points
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution0.1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min ISE0 to 19,990 ppm
  • Min Conductivity0.00 to 3000 mS
  • Min TDS0 to 19,999 ppm
  • TDS Resolution1 ppm
  • Min Salinity0 to 80.0 ppt NaCl equivalent
  • Min Resistivity0.0001 to 100 MΩ
  • Resistivity Accuracy0.5% ±1 digit
  • Min Dissolved Oxygen0.00 to 90 ppm, 0 to 600%
  • Temperature compensationAutomatic or manual
  • WaterproofYes
  • Width (cm)9.525
  • Height (cm)4.445
  • Width (in)3 3/4
  • Height (in)1 3/4
  • DescriptionOptional pH/ISE Meter
  • Warranty3 year
3 Year WarrantyConformite EuropeenneCanadian Standards AssociationIP 67UL Listed.

Key Features

  • Improved stability algorithm increases calibration and measurement accuracy
  • Ensure temperature accuracy by performing the ATC temperature calibration
  • Increased memory allows you to log more data points or turn off the data log if desired
  • Print or view your last 10 calibrations
  • New individual method passwords provide better security
  • 3 year warranty

More About this Item

Star series portable meters feature simple three-button operation with convenient Quick-Start guides to help you get started and provide support while in use. Smart Stability™ and Smart Averaging™ automatically optimize the measurement process to ensure accurate results. Meters offer a full complement of GLP-related features including datalogging with time/date stamp, electrode verification, and optional 21 CFR Part 11 software.

Meters include: Quick-Start guide and four AA batteries.

Meter kits add: waterproof Triode™ pH/ATC electrode; single-use pouches of pH 4.01, 7.00, and 10.01 buffers; pH storage solution; and hard carrying case.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat