Thermo Scientific Precision 096452 Fog Tester Reflectometric Accessory Kit
Representative image only.
Thermo Scientific Precision Mfr # 096452 – Item # EW-24100-07
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionFog Tester Reflectometric Accessory Kit
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
A Test Method Kit and bath fluid are the third components needed to create a Horizon Fog Testing System. Choose a primary bath, cooling system and one, or both, test method start-up kits to complete a system that meets your needs.
Kits contain everything needed for testing; order heat transfer fluid separately (required).
Reflectrometric Kit: 6 cooling plates, 6 glass beakers, 6 metal rings, 6 elastomer sealing rings, 6 support rings, 6 square glass plates, 1 frame for glass plates, 1 sample cutter, 1 set of covers and 1 refractometer.