Thermo Scientific Precision TSDUB15 Shaking Water Baths, +/-0.1C, 15L Dubnoff

Representative image only.
Thermo Scientific Precision Mfr # TSDUB15 – Item # EW-12150-03
Easily clean and maintain bath with coil-free design
  • Oscillations per Min: 0, 30-200
  • Temperature range: 5 C above ambient to 100 C
  • Temperature uniformity at 70 C: ± 0.05 C
  • Temperture stability at 70 C: ± 0.1 C
  • 2 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Bath TypeShaking
  • Reservoir Capacity (L)15
  • Max Temperature (° C)100
  • Min Temperature (° C)Ambient +5
  • Temperature Uniformity (° C)±0.05
  • Width (in)15 1/2
  • Length (in)36 29/32
  • Height (in)9 51/64
  • Bath Opening Width (in)12
  • Bath Opening Length (in)11 1/2
  • Immersion Depth (in)12
  • Heater Wattage (watts)1200
  • Power (VAC)115 / 230
  • Power (Hz)50/60
  • DescriptionShaking Water Baths, +/-0.1C, 15L Dubnoff
  • Warranty2 year
Two year warranty

Key Features

  • Oscillations per Min: 0, 30-200
  • Temperature range: 5 C above ambient to 100 C
  • Temperature uniformity at 70 C: ± 0.05 C
  • Temperture stability at 70 C: ± 0.1 C
  • 2 year warranty

More About this Item

Thermo Scientific Precision shaking water baths support a range of sensitive life science and QA/QC applications, from warming fragile reagents to tissue culturing and genetics sequencing. Easily clean and maintain your bath with new coil-free interior. Precision shaking water baths are available in 15 L and 27 L capacities.

Precision shaking water baths are available in 15 L and 27 L capacities. The shallow form model has a capacity of 15 L with a removable tray that provides a depth of 3.5” (8.9 cm) for use with smaller sample containers. The Dubnoff water bath is designed specifically for applications that require your samples to be incubated in a controlled atmosphere; equipped with one large and two small gassing hoods.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat