Thermo Scientific R01580 Mannitol salt agar (MSA), 10/pk
Representative image only.
Thermo Scientific Mfr # R01580 – Item # EW-14206-96
Stringent quality control ensures high-quality performance as intended!
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
PRODUCT OPTIONS
$84.00USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Media Name (No.)Mannitol Salt
  • DescriptionMannitol salt agar (MSA), 10/pk
Key Features
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
More About this Item
Choose from a variety of media formulations. Use this culture media for the isolation and differentiation of microorganisms from samples or specimens. Each raw component is pretested before being used, for quality assurance and consistency from lot batch. Intended for lab practice only; not intended for in vitro purposes. Plates have stacking rings which interlock to minimize sliding.