Thermo Scientific R08652 Prepared Media Tube, Potato Dex Agar Slant 20/pk
Representative image only.
Thermo Scientific Mfr # R08652 – Item # EW-14207-58
Provide fast and efficient quality results!
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
SPECS & DESCRIPTION
Specifications & Description
  • Media Name (No.)Potato Dextrose
  • Tube Length (mm)113
  • Tube Diameter (mm)20
  • DescriptionPrepared Media Tube, Potato Dex Agar Slant 20/pk
Key Features
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
More About this Item
Choose from a variety of media formulations. Use this culture media for the isolation and differentiation of microorganisms from samples or specimens. Most are for immediate use — no further preparation is necessary. Each raw component is pretested before being used, for quality assurance and consistency from lot batch.