Thermo Scientific R112022 Prepared Bottles, Brain Heart Inf (BHI) Agar, 500 mL; 10/Pk
Representative image only.
Thermo Scientific Mfr # R112022 – Item # EW-14208-04
Ensure the quality of your sample
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
SPECS & DESCRIPTION
Specifications & Description
  • Media Name (No.)Brain-Heart Infusion
  • Capacity (mL)500
  • Capacity (L)0.5
  • DescriptionPrepared Bottles, Brain Heart Inf (BHI) Agar, 500 mL; 10/Pk
Key Features
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
More About this Item
Choose these prepared bottles for your media formulation. Use this culture media for the growth and dilution of microorganisms from samples. Ready for immediate use; no further preparation is necessary. Each raw component is pretested for quality assurance and consistency from each lot batch.