Thermo Scientific R452602 Dehydrated Culture Media, Brilliant Green Bile broth, 500 g
Representative image only.
Thermo Scientific Mfr # R452602 – Item # EW-14206-10
Stringent quality control ensures reliable performance
  • For lab use only, not intended for in vitro purposes
SPECS & DESCRIPTION
Specifications & Description
  • Media Name (No.)Brilliant Green Bile
  • Capacity (g)500
  • DescriptionDehydrated Culture Media, Brilliant Green Bile broth, 500 g
Key Features
  • For lab use only, not intended for in vitro purposes
More About this Item
Use this culture media for isolation and differentiation of microorganisms from samples or specimens. Choose from a variety of media formulations. Each raw component is pretested before being used for quality assurance and consistency from lot batch. All animal product components are in compliance with USDA and APHIS regulations. For lab use only, not intended for in vitro purposes.
For the detection of coliform bacteria in water, food, and dairy products.
Available for sale in the United States only