Thermo Scientific R453942 Dehydrated Culture Media, m ENDO agar LES, 500 g
Representative image only.
Thermo Scientific Mfr # R453942 – Item # EW-14206-06
Stringent quality control ensures reliable performance
  • For lab use only, not intended for in vitro purposes
PRODUCT OPTIONS
$463.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Media Name (No.)m-ENDO
  • Capacity (g)500
  • DescriptionDehydrated Culture Media, m ENDO agar LES, 500 g
Key Features
  • For lab use only, not intended for in vitro purposes
More About this Item
Use this culture media for isolation and differentiation of microorganisms from samples or specimens. Choose from a variety of media formulations. Each raw component is pretested before being used for quality assurance and consistency from lot batch. All animal product components are in compliance with USDA and APHIS regulations. For lab use only, not intended for in vitro purposes.
For the enumeration of coliforms by membrane filtration.
Available for sale in the United States only