Thermo Scientific R454182 Dehydrated Culture Media, Nutrient agar, 500 g
Representative image only.
Thermo Scientific Mfr # R454182 – Item # EW-14206-78
Stringent quality control ensures reliable performance
  • For lab use only, not intended for in vitro purposes
SPECS & DESCRIPTION
Specifications & Description
  • Media Name (No.)Nutrient
  • Capacity (g)500
  • DescriptionDehydrated Culture Media, Nutrient agar, 500 g
Key Features
  • For lab use only, not intended for in vitro purposes
More About this Item
Use this culture media for isolation and differentiation of microorganisms from samples or specimens. Choose from a variety of media formulations. Each raw component is pretested before being used for quality assurance and consistency from lot batch. All animal product components are in compliance with USDA and APHIS regulations. For lab use only, not intended for in vitro purposes.
General-purpose medium for cultivation of less fastidious microorganisms.
Available for sale in the United States only