Thermo Scientific REL-1204D Laboratory space-saver refrigerator (single door); 11.5 CU FT; 200/230V
Representative image only.
Thermo Scientific Mfr # REL1204D – Item # EW-44185-15
Maintain uniform temperature stability—even with sporadic door openings!
  • Two-digit LED temperature display
  • Constructed of heavy0gauge steel
  • Audible and visual alarm with high and low set points
  • 13 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Freezer Capacity (cu ft)17.2
  • Freezer Capacity (Liters)487
  • Min Temperature - Freezer (° F)34
  • Max Temperature - Freezer (° F)46
  • Min Temperature - Freezer (° C)1
  • Max Temperature - Freezer (° C)8
  • Temperature ControlMicroprocessor
  • Number of Doors1
  • Number of Shelves5
  • AlarmYes
  • DisplayLED
  • Power (VAC)208 / 230
  • Power (Hz)60
  • Power (amps)12
  • Power (Breaker)15 A
  • DescriptionLaboratory space-saver refrigerator (single door); 11.5 CU FT; 200/230V
  • Warranty13 month
Conformite EuropeenneCanadian Standards AssociationUL Listed.
Key Features
  • Two-digit LED temperature display
  • Constructed of heavy0gauge steel
  • Audible and visual alarm with high and low set points
  • 13 month warranty
More About this Item
Exclusive IntrLogic™ control simplifies operation and improves temperature consistency. These refrigerators feature heavy-duty, hermetically sealed compressors with extra sound insulation to minimize noise. Compressors use environmentally safe CFC-free refrigerant. Positive, forced-air circulation maintains temperature uniformity at all shelf levels.
Key-operated on/off switch gives added protection against tampering. Audible and visual alarms alert in case of open door, low battery, system maintenance, or power failure. The refrigeratorsalso feature zinc-plated open wire shelves that are
adjustable in 1" increments.