Thermo Scientific Revco UEN-2320A Enzyme freezer, -20°C, 115 VAC, 60 Hz

Representative image only.
Thermo Scientific Revco Mfr # UEN-2320A – Item # EW-44164-00
Microprocessor driven for precise control and documentation
  • Enzyme freezer offers organized inventory system
  • Interlogic™ microprocessor control system maintains temperature
  • 13 month warranty

SPECS & DESCRIPTION

Specifications & Description

  • Freezer Capacity (cu ft)23.3
  • Freezer Capacity (Liters)659
  • Min Temperature - Freezer (° F)-4
  • Min Temperature - Freezer (° C)-20
  • Temperature Accuracy (° C)±1
  • Temperature ControlMicroprocessor
  • Freezer Interior Width (in)24
  • Freezer Interior Height (in)58
  • Freezer Interior Depth (in)29
  • Freezer Interior Width (cm)61
  • Freezer Interior Height (cm)147
  • Freezer Interior Depth (cm)74
  • Exterior Width (in)28
  • Exterior Height (in)77 1/2
  • Exterior Depth (in)35 1/2
  • Exterior Width (cm)71
  • Exterior Height (cm)197
  • Exterior Depth (cm)90
  • Number of Doors1
  • Number of Shelves9
  • Number of Boxes45
  • AlarmAudible/visual
  • Cabinet ConstructionHeavy-gauge steel
  • DisplayDigital
  • Power (VAC)115
  • Power (Hz)60
  • Power (amps)13.5
  • Power (Breaker)20
  • DescriptionEnzyme freezer, -20°C, 115 VAC, 60 Hz
  • Warranty13 month
UL Listed to Canadian safety standards.UL Listed.

Key Features

  • Enzyme freezer offers organized inventory system
  • Interlogic™ microprocessor control system maintains temperature
  • 13 month warranty

More About this Item

Check operating status at a glance with enlarged digital display and graphic thermometer. Set temperature and alarm set points in 1°C increments. IntrLogic™ microprocessor control provides better accuracy than other freezers. Set point security system protects your samples with a key-operated switch for main power and alarm locks in temperature and alarm set points. Alarm system audibly and visually identifies any temperature deviation beyond alarm set points and archives deviated temperatures. Extreme ambient alert warns if room temperature may have affected performance.

Gravity convection air system provides maximum temperature stability without blowers or fans. Manual defrost avoids intermittent coil warming during auto defrost cycles. Each model is constructed of heavy gauge steel and features hermetically sealed compressors and a single self-closing solid door. The full length door handle has keyed lock for security.

Includes an organized inventory system—nine shelves hold 45 bins (five bins per shelf), making them ideal for enzyme storage. Adjust shelf height in 1-inch increments without tools.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat