Thermo Scientific SPEEDVAC Savant SpeedVac Kit OP115V

Representative image only.
Thermo Scientific Mfr # DNA120OP-115 – Item # EW-13045-10
Everything you need under one catalog number --eliminating guess work!
  • Rapid and efficient concentration of small-volume samples
  • Extremely quiet operation for the laboratory
  • Packages come complete with main components: concentrator, pump, rotor, and trap
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Evaporation Flask Size1.5mL
  • Tubes Held36
  • Timer (min)Yes
  • Power (VAC)115
  • Power (Hz)60
  • Length (in)11
  • Width (in)11
  • Height (in)25
  • Length (cm)27.94
  • Width (cm)27.94
  • Height (cm)63.5
  • DescriptionSavant SpeedVac Kit OP115V
  • Warranty1 year
1 Year Warranty

Key Features

  • Rapid and efficient concentration of small-volume samples
  • Extremely quiet operation for the laboratory
  • Packages come complete with main components: concentrator, pump, rotor, and trap
  • 1 year warranty

More About this Item

Easy-to-use, small-capacity systems are the ideal choice for concentrating or drying DNA/RNA samples. The PTFE-coated chamber is corrosion resistant for lasting service. Dual controls allow you to set independent heat and run times, with choice of three drying times.The systems are designed to accommodate microcentrifuge tubes. A built-in, oil-free PTFE pump produces the vacuum required for rapid drying with no regular maintenance required.

Savant DNA120-OP package is designed for use with oligos; it features a glass cover that is chemically resistant to ammonia hydroxide. Recommended solvents include PCR buffers, isopropanol, and ammonium hydroxide. The system includes a DNA concentrator with integral pump, RD36 rotor for 36 x 1.5 mL microtubes, ANT100 chemical trap, and ANS121 ammonium neutralizing solution.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat