Thermo Scientific STAR A215 Star A215 pH/Conductivity Benchtop Meter Kit

Representative image only.
Thermo Scientific Mfr # STARA2155 – Item # EW-58825-38
Durability and dependability make it the ideal choice for every lab, every sample, everyday
  • Multilanguage interface supports English, Spanish, French, Italian, German and Chinese
  • Improved stability algorithm provide better measurement accuracy
  • Meets US Pharmacopoeia (USP) requirement
  • Ensure temperature accuracy with ATC temperature calibration
  • Nonvolatile memory holds up to 2000 data points with time date stamp

PRODUCT OPTIONS

$2,810.00USD / Each

$178.00 - $185.00USD / Each

for an additional
$178.00USD / Each
for an additional
$185.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeBenchtop
  • Display typeGraphic LCD
  • ConnectionspH: BNC; Temperature: 8-pin mini-DIN
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.002 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 5 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.01 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min TDS0 ppt
  • Max TDS200 ppt
  • Min Salinity0 ppt
  • Max Salinity80 ppt
  • Resistivity Resolution2 Ω
  • Resistivity Accuracy±0.5% ±1 digit
  • Dissolved Oxygen Accuracy±0.5% of reading
  • Min Temperature (° C)-5
  • Max Temperature (° C)105
  • Min Temperature (° F)23
  • Max Temperature (° F)221
  • InterfaceRS-232 and USB
  • WaterproofYes
  • Power (VAC)100 to 240
  • Power (Hz)50 or 60
  • Battery4 x AA batteries (optional)
  • Width (cm)17.78
  • Height (cm)24.765
  • Width (in)7
  • Length (in)3
  • Height (in)9 3/4
  • DescriptionStar A215 pH/Conductivity Benchtop Meter Kit

Key Features

  • Multilanguage interface supports English, Spanish, French, Italian, German and Chinese
  • Improved stability algorithm provide better measurement accuracy
  • Meets US Pharmacopoeia (USP) requirement
  • Ensure temperature accuracy with ATC temperature calibration
  • Nonvolatile memory holds up to 2000 data points with time date stamp
  • Individual method passwords provide better security
  • Option of AC power or four AA batteries (sold separtely)

More About this Item

Thermo Scientific Orion Star A215 meter offers pH, mV, RmV, conductivity, salinity, TDS, resistivity and temperatur. Meter features an intuitive user interface with plain language prompts. A large, backlit graphic LCD shows measurements with temperature along with electrode status, time/date, sample ID, user ID, and calibration data.

GLP-related features include 2000 data points with time/date stamp and electrode verification. Meter utilizes RS-232 or USB for connectivity and includes data analysis software.

Up to five point automatic pH calibration with USA/NIST and Din buffers, auto-buffer recognition, and recalibration feature to fix calibration errors.

Selectable cell constant for use with 2- or 4- band conductivity probes and selectable reference temperatures of 5, 10, 15, 20, 25°C ensure the most accrate conductivity measurement.

Available as meter only and as complete meter kit configurations.

Meter kit includes: ROSS™ Triode pH/ATC electrode (05718-28), DuraProbe conductivity cell (19704-70), 475-ml of each 4, 7, 10 buffer and storage solution, 5 x 60-ml of 1413µS/cm standard, swing-arm electrode holder, and universal power adapter.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-847-549-7600
Live chat