Thermo Scientific TSG General Purpose Chromatography Refrigerator, 27 cuft, Gray Exterior, Glass Door; 115 VAC

Representative image only.
Thermo Scientific Mfr # TSG30CSGA – Item # EW-44261-37
Go green while improving cold storage
  • EPA ENERGY STAR® certified high-efficiency cooling system
  • Green, natural refrigerants offer greater safety
  • Whisper-quiet compressor and low heat output fits in any lab setting
  • Temperature high and low, door ajar, and power failure alarms protect critical supplies and samples
  • Protect samples with the safety sensor interior outlet
  • Feature a painted gray exterior and stainless steel interior, 27 cubic foot capacity, and a single glass door

SPECS & DESCRIPTION

Specifications & Description

  • Refrigerator Capacity (cu ft)27
  • Refrigerator Capacity (Liters)764
  • Min Temperature - Refrigerator (° C)2
  • Max Temperature - Refrigerator (° C)10
  • Temperature ControlDigital
  • Refrigerator Interior Width (in)28 1/2
  • Refrigerator Interior Height (in)27
  • Refrigerator Interior Depth (in)61 3/4
  • Refrigerator Interior Width (cm)72.4
  • Refrigerator Interior Height (cm)68.6
  • Refrigerator Interior Depth (cm)157
  • Exterior Width (in)32
  • Exterior Height (in)30 3/4
  • Exterior Depth (in)81
  • Exterior Width (cm)81.3
  • Exterior Height (cm)78
  • Exterior Depth (cm)205.7
  • Number of Doors1
  • Number of Shelves4 full, 3 half
  • AlarmYes
  • Cabinet ConstructionStainless Steel
  • Power (VAC)115
  • Power (Hz)60
  • Power (amps)3
  • DescriptionGeneral Purpose Chromatography Refrigerator, 27 cuft, Gray Exterior, Glass Door; 115 VAC
Two year warrantyUL Listed to Canadian safety standards.

Includes

adjustable shelves and half shelves, shelving clips, casters, wrench, door lock key, power key switch, and power cord.

Key Features

  • EPA ENERGY STAR® certified high-efficiency cooling system
  • Green, natural refrigerants offer greater safety
  • Whisper-quiet compressor and low heat output fits in any lab setting
  • Temperature high and low, door ajar, and power failure alarms protect critical supplies and samples
  • Protect samples with the safety sensor interior outlet
  • Feature a painted gray exterior and stainless steel interior, 27 cubic foot capacity, and a single glass door

More About this Item

Ideal for storing reagents, media, and samples, these general purpose chromatography refrigerators are also engineered to be better for the environment. Save electricity and money with adaptive defrost, decreased compressor cycle time, and reduced heat transfer. The natural refrigerant R290 will not deplete the ozone layer or contribute substantially to climate change. Being manufactured in a zero-waste-to-landfill facility ensures at least 99% of the waste produced is reused, recycled, composted, incinerated, or used in energy recovery.

The refrigerators’ quiet 51-dBA operation and low heat output allows for installation near workstations for added convenience. Visible and audible alarms alert you when temperature exceeds high or low limits and when there is a power outage prevent losses of valuable contents. Durable steel construction withstands chemical spills and resists corrosion.

The chromatography refrigerators are equipped with a hydrocarbon safety circuit sensor to ensure safety. In the event the safety circuit is tripped and the safety sensor senses a flammable gas, the power to the internal outlet will be discontinued.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat