Thermo Scientific TSX High-Performance Undercounter Refrigerator, 5.5 cuft, Glass Door; 100-230 VAC, UK Plug

Representative image only.
Thermo Scientific Mfr # TSX505GW – Item # EW-44202-09
Redundant cooling technology uses 37% less energy, emits 20X less noise and delivers twice the storage capacity
  • Variable speed control uses thermoelectric devices instead of compressors providing ideal conditions for most applications
  • Whisper-quiet operation around 35 dBA allows use in work station areas
  • Removal of compressor allows for twice the storage capacity of similar size units
  • 2 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Refrigerator Capacity (cu ft)5.5
  • Refrigerator Capacity (Liters)156
  • Min Temperature - Refrigerator (° F)35.6
  • Max Temperature - Refrigerator (° F)46.4
  • Min Temperature - Refrigerator (° C)2
  • Max Temperature - Refrigerator (° C)8
  • Temperature ControlIcon Based User Interface
  • Refrigerator Interior Width (in)19 1/2
  • Refrigerator Interior Height (in)23
  • Refrigerator Interior Depth (in)20 1/2
  • Refrigerator Interior Width (cm)49.6
  • Refrigerator Interior Height (cm)58.5
  • Refrigerator Interior Depth (cm)52.1
  • Exterior Width (in)23 39/64
  • Exterior Height (in)31 13/16
  • Exterior Depth (in)26
  • Exterior Width (cm)60
  • Exterior Height (cm)80.8
  • Exterior Depth (cm)66.1
  • Number of Doors1 Glass
  • Number of Shelves3
  • Number of Boxes495
  • AlarmAudible and Visual
  • Cabinet ConstructionPainted steel with interior aluminum
  • DisplayFull Color LCD
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • Power (amps)5
  • Description5.5 cu ft, Glass Door, 100-230 VAC, UK Plug
  • Warranty2 year
Two year warranty

Key Features

  • Variable speed control uses thermoelectric devices instead of compressors providing ideal conditions for most applications
  • Whisper-quiet operation around 35 dBA allows use in work station areas
  • Removal of compressor allows for twice the storage capacity of similar size units
  • 2 year warranty

More About this Item

Designed for a demanding lab environment the TSX Series undercounter refrigerator provides excellent cold storage of samples, media, and reagents in a compact space. Unique compressor-free V-drive technology provides reduced energy consumption and quiet operation without compromising performance. The cooling system actively monitors the cabinet to maintain the desired temperature conditions. This constant monitoring and adjustment keeps the temperature stable without the fluctuations found with compressors that cycle on and off, or with periods of door openings. The tight internal air circulation eliminates the risk of freezing which can ruin valuable samples. A large 4.5" full color TFT-LCD user interface makes operation simple and easy. The icon based touchscreen controller features all the parameters for operation and includes password protection, adjustable lighting, built-in datalogging, USB port, and an ergonomic tilted design. Audible and visual alarms alert user to power fail, high temp, low, temp, low battery memory full, and door ajar. Removal of compressor allows for almost 2-times more storage than units of similar size and footprint.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat