Thermo Scientific TX-750 Swinging Bucket Rotor; 4 x 750 mL
Representative image only.
Thermo Scientific Mfr # 75003180 – Item # EW-17708-00
  • Rotor body only with "Slide Coat" anti-friction finish — buckets, lids, and any needed adapters should be ordered separately
PRODUCT OPTIONS
$3,138.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Speed (rpm)4700
  • Max RCF x g4816
  • Rotor TypeSwing-out
  • Tube Sizes Accepted750 mL
  • Number Of Tubes Accepted4
  • DescriptionSwingingBucketRotor;4x750mL
Key Features
  • Rotor body only with "Slide Coat" anti-friction finish — buckets, lids, and any needed adapters should be ordered separately
More About this Item
Choose the rotor compatible with your Thermo Scientific centrifuge and application needs.
Compatible with: Thermo Scientific Sorvall™ ST40/ST40R, XT/XTR/XF/XFR, Heraeus™ Megafuge™ 40/40R, Multifuge™ X3/X3R, and Thermo Scientific SL40/40R