ThermoSafe 304 Dry Ice Storage Chest, tabletop, polyethylene, 50 lb capacity
Representative image only.
ThermoSafe Mfr # 304 – Item # EW-03719-00
Holds up to 50 lb of dry ice
$242.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lbs)50
  • DescriptionDry Ice Storage Chest, tabletop, polyethylene, 50 lb capacity
More About this Item
Tabletop chest has dry ice sublimation rate less than 2.5 lb per day at full 50 lb capacity. Chest is constructed from rugged polyethylene; molded bottom won't scratch tabletop. Inside dimensions are 11-1/2"L x 9-3/4"W x 9-1/4"H.