ThermoSafe 435 Brands Aluminum Can Shipper, 48/cs

Representative image only.
ThermoSafe Mfr # 435 – Item # EW-03727-04
Ship non-hazardous specimens safely
  • Fitted halves cushion tubes for superior protection
  • Lightweight mailers meet postal regulations for nonhazardous materials

SPECS & DESCRIPTION

Specifications & Description

  • Inside Height (in)4 3/4
  • Inside Diameter (ID) (in)1 3/4
  • Inside Height (cm)12.1
  • Inside Diameter (ID) (cm)4.4
  • Shipping Weight (oz)35.2
  • Shipping Weight (g)998
  • DescriptionAluminum Shipping Can Shipper, 48/cs

Key Features

  • Fitted halves cushion tubes for superior protection
  • Lightweight mailers meet postal regulations for nonhazardous materials

More About this Item

Protect your fragile samples with this expanded polystyrene shipper. Light weight minimizes postage costs. Strong white paperboard shipping carton (sold separately) has sure-lock tabs that eliminate taping or tying. Choose from tube mailers, slide mailer, mini mailer, multipurpose mailer, or aluminum shipping can.

Accomodates most commonly used tubes, vials, and bottles. Provides additional security during handling and shipping. Screw-cap and seamless construction prevents leakage. Completely autoclavable.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat