Todd Systems SD-16 G Inc Step-down transformer, 100 Watts
Representative image only.
Todd Systems Mfr # SD-16 G – Item # EW-01578-02
Power your instruments with 110/120 or 220/240 VAC outlets
  • Step-up models include NEMA 6-15P 230 VAC adapter for wiring convenience
  • Include rugged NEMA cord with three-prong plug
PRODUCT OPTIONS
$60.35USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Input (VAC)220240
  • Output (VAC)110/120
  • Power Rating (watts)100
  • DescriptionInc Step-down transformer, 100 Watts
Key Features
  • Step-up models include NEMA 6-15P 230 VAC adapter for wiring convenience
  • Include rugged NEMA cord with three-prong plug
More About this Item
Select a step-down transformer to connect 110/120 VAC instruments to 220/240 VAC power sources; use a step-up transformer to connect 220/240 VAC instruments to 110/120 VAC power sources. All models run on 50/60 Hz.