Torque Wrench, 3/8in Drive, 30/250in.LB Micro-adj Metal Handle
Representative image only.
Item # EW-97610-01
  • FEATURES
    • Dual scale, calibrated dual direction
    • Positive lock with spring loaded pull down lock ring
    • Fast, accurate and easy to set
    • Easy to read laser marked scales
    • Most feature Quick Release buttons which provide good socket retention as well as easy socket removal
    • Accuracy: ± 4% clockwise ± 6% CCW of indicated value, CW, from 20 to 100% of full scale
    • Meets or exceeds ASME B107.14M-1994, ISO 6789 standards
    • Individually serialized with matching certificate of calibration traceable to NIST
PRODUCT OPTIONS
$343.25USD / Each
$138.00 - $211.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lb-in)250
  • Capacity (N-m)27.7
  • Drive (in)0.375
  • Length (in)11.25
  • Capacity250 in lb / 27.7 N m
  • DescriptionTorque Wrench, 3/8in Drive, 30/250in.LB Micro-adj Metal Handle
Key Features
  • FEATURES
    • Dual scale, calibrated dual direction
    • Positive lock with spring loaded pull down lock ring
    • Fast, accurate and easy to set
    • Easy to read laser marked scales
    • Most feature Quick Release buttons which provide good socket retention as well as easy socket removal
    • Accuracy: ± 4% clockwise ± 6% CCW of indicated value, CW, from 20 to 100% of full scale
    • Meets or exceeds ASME B107.14M-1994, ISO 6789 standards
    • Individually serialized with matching certificate of calibration traceable to NIST
More About this Item
Micro-adjustable torque wrenches; metal handle English scale
The metal handle series offers the user a rugged, industrial strength torque wrench able to withstand the rigors of professional use. handles are lightly knurled for a tough, non-slip grip. There are 34 models of metal handle micrometer adjustable torque wrenches available