Total Source APP0242-W Polyethylene-Coated Polypropylene Lab Coats, X-large, case of 30
Representative image only.
Total Source Mfr # APP0242-W-XL – Item # EW-86227-16
Excellent lightweight splash protection
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)41.5
  • Length (cm)105.41
  • SizeX-Large
  • MaterialPolyethylene(PE)
  • ColorWhite
  • DescriptionPolyethylene-Coated Polypropylene Lab Coats, X-large
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
These polyethylene (PE) coated polypropylene (PP) lab coats are a low-cost disposable alternative to Tyvek® coats. The durable, lightweight, and breathable fabric is ideal for general-purpose laboratory applications. The PE-coated spun bond PP-provides excellent splash protection from water-based liquids.