Total Source APP0380-21-18 Polyethylene (PE)-Coated Polypropylene (PP) Sleeves, 18" Length
Representative image only.
Total Source Mfr # APP0380-21-18 – Item # EW-86227-70
Splash protection in a comfortable, breathable material
  • Ideal for general-purpose laboratory use
PRODUCT OPTIONS
$94.00USD / Case of 200
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)18
  • Length (cm)45.72
  • MaterialPolyethylene
  • ColorWhite
  • DescriptionPolyethylene (PE)-Coated Polypropylene (PP) Sleeves, 18" Length
Key Features
  • Ideal for general-purpose laboratory use
More About this Item
These polyethylene-coated polypropylene sleeves are a low cost, disposable alternative to Tyvek®. The fabric is durable, lightweight, and breathable. The PE-coated spun bond PP provides excellent splash protection from water based liquids.
Call Us At1-847-549-7600
Live chat