TPI 605 Digital Vacuum Gauge, 12000 to 15 micron
Representative image only.
TPI Mfr # 605 – Item # EW-42500-18
Verify vacuum before you charge a refrigerant system
  • Measure vacuum from 25 to 12,000 microns
  • Reads vacuum in seven international units of measure (microns, PSI, InHg, mBar, Pascals, Torr, and mTorr)
  • Display updates every 0.5 seconds
  • Auto power off after 10 minutes when vacuum above 12,000 microns (12 Torr)
  • Built in hook for hanging
$129.75USD / Each
$146.00 - $216.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±0.5% of reading + 5 digits
  • Max Vacuum (" Hg)30
  • Battery Type9 V
  • DescriptionDigital Vacuum Gauge, 12000 to 15 micron
  • Warranty3 year
3 Year Warranty
Key Features
  • Measure vacuum from 25 to 12,000 microns
  • Reads vacuum in seven international units of measure (microns, PSI, InHg, mBar, Pascals, Torr, and mTorr)
  • Display updates every 0.5 seconds
  • Auto power off after 10 minutes when vacuum above 12,000 microns (12 Torr)
  • Built in hook for hanging
  • Clean sensor with alcohol
  • 500 psi overpressure
  • 3 year warranty
More About this Item
Prevents moisture and air from remaining in the sytem. Finds small leaks.