TPI Digital Manometer, +/-80.29 in. H2O, Single Input

Representative image only.
TPI Mfr # 610 – Item # EW-42500-19
Replace messy U-tubes (water column manometers) and expensive pressure gauges with digital manometers
  • Measure pressure of air and natural gas Auto power off after 40 minutes
  • 3 year warranty

PRODUCT OPTIONS

$164.00USD / Each

$152.00 - $225.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Accuracy±0.5% of reading + 5 digits
  • Min Pressure (in WC)-15
  • Max Pressure (in WC)48
  • Min Temperature (° F)32
  • Max Temperature (° F)104
  • Min Temperature (° C)0
  • Max Temperature (° C)60
  • Process Connection1/4\" hose barb
  • Unit Of Measurement\" WC
  • Display4-digit LCD
  • Battery Type1 9V (included)
  • Media CompatibilityAir and natural gas
  • Resolution0.001\" WC
  • DescriptionDigital Manometer, +/-80.29 in. H2O, Single Input
  • Warranty3 year
3 Year Warranty

Includes

one 9V battery.

Key Features

  • Measure pressure of air and natural gas Auto power off after 40 minutes
  • 3 year warranty

More About this Item

Applications include adjusting appliance regulators, setting delivery pressures, leakage "shut-in" tests, regulator adjustments for pressure and lock-up tests, tests for leaks in low-pressure gas systems , isolate difficult pressure problems on natural gas and propane systems, test pressure switch operation on a 2-stage gas furnace in both positive and negative pressures, static pressures in air ducts for air balancing, and checking air filters by measuring pressure drop

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat