TPI 621 Dual Input Differential Manometer, +/-5 psi
Representative image only.
TPI Mfr # 621 – Item # EW-42500-12
$200.00USD / Each
$164.00USD / Each
for an additional
$164.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±0.5% of reading 5 digits
  • Max Pressure (PSI)5
  • Min Pressure (in WC)-120
  • Max Pressure (in WC)120
  • Min Temperature (° F)32
  • Max Temperature (° F)104
  • Min Temperature (° C)0
  • Max Temperature (° C)60
  • Process Connection1/4" hose barb
  • Unit Of Measurementmm Hg, " Hg, kPa, mBar, psi, " WC, mm WC
  • Display4-digit LCD
  • Battery Type9 V (included)
  • Media CompatibilityAir and natural gas
  • Resolution0.001" WC
  • DescriptionDual Input Differential Manometer, /-5 psi
Two year warranty
More About this Item
Replace messy U-tubes (water column manometers) and expensive pressure gauges with digital manometers.
FEATURES
  • Measure pressure of air and natural gas
  • Auto Power Off after 40 minutes
      APPLICATIONS
      • Adjusting appliance regulators
      • Setting delivery pressures
      • Leakage "shut-in" test
      • Regulator adjustments for pressure and lock-up tests
      • Test for leaks in low-pressure gas systems
      • Isolate difficult pressure problems on natural gas and propane systems
      • Test pressure switch operation on a 2-stage gas furnace in both positive and negative pressures
      • Static pressures in air ducts for air balancing
      • Check air filters by measuring pressure drop