TPI 775 Ambient CO/Combustion Gas Detector
Representative image only.
TPI Mfr # 775 – Item # EW-10104-25
$375.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Gas MeasuredCarbon Monoxide
  • DescriptionAmbient CO/Combustion Gas Detector
More About this Item
The TP775 is a state-of-the-art carbon monoxide and combustible gas leak detector. It is the first of its kind to combine both detection technologies into one easy to use hand-held affordable instrument.
FEATURES
    Digital readout for CO and leak level indication
  • One-handed operation
  • Auto-zero
  • Backlight
  • Audible and visual alarms
  • Flexible 16" gooseneck
  • Soft carrying case