TPI Carbon Monoxide Analyzer Kit; 0 to 10,000 ppm
Representative image only.
TPI Mfr # 707 – Item # EW-10104-02
Choose from a complete line of flue gas and CO analyzers
  • When selecting oil as fuel be sure to use the optional oil filter TP773 or reading could become erratic. You must use the TP715K3 kit which includes the TP784 regulator when working on oil and gas pipeline diesel compression stations The TP784 flow regulator must be used in any application where high velocity pressure is present
  • 3 year warranty
PRODUCT OPTIONS
$433.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Min CO Range (ppm)0
  • Max CO Range (ppm)10000
  • Memory10 readings
  • Battery Type3 AA (not included)
  • DescriptionCarbon Monoxide Analyzer Kit; 0 to 10,000 ppm
  • Warranty3 year
3 Year Warranty
Key Features
  • When selecting oil as fuel be sure to use the optional oil filter TP773 or reading could become erratic. You must use the TP715K3 kit which includes the TP784 regulator when working on oil and gas pipeline diesel compression stations The TP784 flow regulator must be used in any application where high velocity pressure is present
  • 3 year warranty
More About this Item
Choose from a complete line of flue gas and CO analyzers. All units measure CO and depending on the model, you can additionally measure O2, temperature, pressure, and NO. Calculate CO 2, gross of net efficiency, excess air and NOX. All models include flue probe and carrying case. Select the model that is right for you.