Traceable Micro Probe for Scientic Thermistor Thermometer 90080-12
Representative image only.
Traceable Item # EW-90080-14
Optimize performance of your thermometer
  • Probe is submersible up to the handle for ideal measuring
  • Features a temperature range of –58 to 302°F (–50 to 150°C)
  • Probe has 0.1 cm diameter, 7 cm stem length, 14 cm overall length, and 152 cm cable length
  • Traceable® model number 4021
PRODUCT OPTIONS
$126.00USD / Each
$165.00 - $183.00USD / Each
for an additional
$165.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Length (mm)70
  • Diameter (mm)1
  • DescriptionMicro Probe for Scientic Thermistor Thermometer 90080-12
Key Features
  • Probe is submersible up to the handle for ideal measuring
  • Features a temperature range of –58 to 302°F (–50 to 150°C)
  • Probe has 0.1 cm diameter, 7 cm stem length, 14 cm overall length, and 152 cm cable length
  • Traceable® model number 4021
More About this Item
Compatible for use with the Traceable Scientific Thermistor Thermometer 90080-12, meter sold separately.