Traceable Multi-Parameter Pen-Style Meter with Calibration
Representative image only.
Traceable Item # EW-37950-04
Portable, lightweight and easy-to-use
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • User selectable dual display shows humidity/temperature, humidity/dew point, barometer/temperature, or barometer/humidity
  • Water-resistant design with O-ring seal and membrane keypad
  • Traceable® model number 4247
$217.00USD / Each
Time to recalibrate?
for an additional
$70.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Min RH Range (%)10
  • Max RH Range (%)95
  • RH Accuracy±3% RH
  • RH Resolution (%)0.1%
  • Min Temperature (° F)32
  • Min Temperature (° C)0
  • Max Temperature (° F)122
  • Max Temperature (° C)50
  • Temperature Accuracy±1.0°C
  • Temperature Resolution0.1°
  • BatteryAAA Battery (4)
  • DescriptionTraceable® Multi-Parameter Pen-Style Meter with Calibration
1 Year WarrantyDEW POINTNIST Traceable Calibration
Includes
NIST-traceable calibration certificate with data.
Key Features
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • User selectable dual display shows humidity/temperature, humidity/dew point, barometer/temperature, or barometer/humidity
  • Water-resistant design with O-ring seal and membrane keypad
  • Traceable® model number 4247
More About this Item
This unique meter can measure temperature, humidity, barometric pressure and dew point. The compact design features an electronic capacitance polymer film sensor not affected by condensation. Tough ABS plastic housing and water-resistant design with O-ring seal and membrane keypad assure a long life in severe lab or harsh plant environments. Features include a hold button that freezes the current reading on the display and a recall button that displays the minimum and maximum readings since the unit was powered on.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Guaranteed to have at least one year of calibration remaining on certificate.