Traceable Calibrated Scientific RTD Probe; Stainless Steel, Handle
Representative image only.
Traceable Item # EW-37804-08
Optimize performance of your thermometer
  • Replacement probe for your Traceable Scientific RTD thermometer
  • Features a handle so probe can be easily held
  • Traceable® model number 6422
  • 1 year warranty
PRODUCT OPTIONS
$356.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeRTD
  • Number Of Probe Channels1
  • Types Of Probes AcceptedRTD
  • Min Temperature (° F)-328
  • Max Temperature (° F)932
  • Min Temperature (° C)-200
  • Max Temperature (° C)500
  • Probe Diameter (in)0.16
  • DescriptionCalibrated Scientific RTD Probe; Stainless Steel, Handle
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Replacement probe for your Traceable Scientific RTD thermometer
  • Features a handle so probe can be easily held
  • Traceable® model number 6422
  • 1 year warranty
More About this Item
Replace your probe or keep a convenient spare.
For improved performance, probes should be calibrated as a system with the meter.