Traceable Temptestr® Super-Mini Infrared Thermometer
Representative image only.
Traceable Item # EW-39642-01
Economical noncontact thermometer fits right in your pocket!
  • Het readings in less than 1 second
  • As-easy-as point, press button, and read
  • Features minimum/maximum memory readings and hold function
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Distance To Target Size Ratio (D:S)1:1
  • EmissivityFixed
  • Min Temperature (° F)-27
  • Max Temperature (° F)230
  • Min Temperature (° C)-33
  • Max Temperature (° C)110
  • Accuracy±2.5% of reading or ±3.6°F (±2.0°C), whichever is greater
  • Resolution0.5°F (0.2°C)
  • Response time<1 second (95% response)
  • Display Type0.5\" LCD
  • Low Battery IndicatorYes
  • BatteryLR44 Battery (2)
  • DescriptionSuper-Mini Temptestr Infrared Thermometer
  • Warranty1 Year
1 Year WarrantyConformite EuropeenneISO9001:2000 Certified Supplier
Key Features
  • Het readings in less than 1 second
  • As-easy-as point, press button, and read
  • Features minimum/maximum memory readings and hold function
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty
More About this Item
This pocket-sized infrared thermometer takes surface temperature readings of any liquid, solid, or semi-solid in less than one second. Operation is simple — turn it on, point at sample, and take reading. Features include switchable °F/°C display, hold function to freeze readings for 7 seconds, continuous monitoring function, adjustable emissivity, minimum/maximum memory readings, flip-open stand, and soft touch keypad.
Noncontact temperature measurements are ideal for food preparation, life sciences, field use, clean rooms, and for fast inspections.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Cole-Parmer provides a wide offering of Professional Infrared Thermometers with varying features and accuracies depending on your application.
We now offer an FDA-cleared infrared thermometer.
Our other infrared thermometers are not approved for medical usage and are not FDA-approved.
For help in selecting an infrared thermometer please contact the Technical Support team at Cole-Parmer.