Traceable Thermohygrometer with Minimum/Maximum Function and Calibration

Representative image only.
Traceable Item # EW-37101-00
MIN/MAX buttons display stored values
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory

$115.00USD / Each
Time to recalibrate?

SPECS & DESCRIPTION

Specifications & Description

  • Min RH Range (%)25
  • Max RH Range (%)95
  • RH Accuracy±5% RH mid-range; ±8% RH otherwise
  • RH Resolution (%)1%
  • Min Temperature (° F)32
  • Min Temperature (° C)0
  • Max Temperature (° F)122
  • Max Temperature (° C)50
  • Temperature Accuracy±2.0°F from 32° to 104°F; ±4.0°F for remaining range (±1.0°C from 0° to 40°C; ±2.0°C for remaining range)
  • Temperature Resolution
  • Temperature Probe TypeThermistor
  • Response Time8 seconds
  • Display Type2-digit LCD, 0.63" H
  • Mounting TypeWall
  • BatteryAAA Battery (2)
  • Height (in)3 3/4
  • Width (in)2 5/8
  • Depth (in)5/8
  • DescriptionTraceable® Thermohygrometer with Minimum/Maximum Function and Calibration
1 Year WarrantyConformite EuropeenneNIST Traceable Calibration

Includes

Hook and loop tape for versatile mounting, bench stand, two AA batteries, and NIST-traceable calibration certificate with data.

Key Features

  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory

More About this Item

Thermohygrometer displays temperature compensated relative humidity in % RH and temperature in °F or °C. Stores minimum and maximum RH and temperature readings — just press the front-panel MIN and MAX buttons to read stored values. Press the RESET button to continue displaying current RH and temperature conditions. Large LCD also indicates ''comfort'', ''wet'', or ''dry'' condition. Thermohygrometer features an integral stand for easy desktop viewing and hook & loop tape for a more versatile placement of the unit.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.

Guaranteed to have at least one year of calibration remaining on certificate.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat