Traceable UVA Light Meter with RS-232 Output and Calibration

Representative image only.
Traceable Item # EW-98766-94
Monitor and keep results through the computer output*
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Recall minimum, maximum, and average readings
  • Features HOLD and zero functions as well as PC output for data acquisition

SPECS & DESCRIPTION

Specifications & Description

  • Accuracy±2% full scale + 2 digits
  • FunctionData hold, minimum, maximum, zero, and percent difference
  • Display TypeLCD
  • Data LoggingNone
  • OutputMicro-USB
  • InterfaceRS-232
  • Intensity Range0.1 µW/cm2 to 19.99 mW/cm2
  • Intensity Resolution0.1 from 0 to 199.9 µW/cm2 , 0.001 from 0.001 to 1.999 mW/cm2, and 0.01 from 0.01 to 19.99 mW/cm2
  • Energy Range0 to 199.9 µW/cm2, 0.200 to 1.999 mW/cm2, and 2.00 to 19.99 mW/cm2 (autoranging)
  • Length (in)7
  • Width (in)3
  • Height (in)1 1/4
  • Power9 V Battery (1)
  • CertificationsNIST-Traceable
  • DescriptionTraceable® UVA Light Meter with RS-232 Output and Calibration
1 Year WarrantyConformite EuropeenneNIST Traceable Calibration

Key Features

  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Recall minimum, maximum, and average readings
  • Features HOLD and zero functions as well as PC output for data acquisition

More About this Item

This UVA (ultraviolet A) light meter measures from 0.1 µW/cm² to 19.99 mW/cm² over 320 to 390 nm. Color-correction filter and precise photodiode produce accurate cosine- and color-corrected measurements over the wide range.

Order the optional data acquisition software to connect to your PC to capture readings. Meter also recalls min, max, and average measurements and features HOLD and Zero functions to freeze the display or obtain a baseline for percent difference.

Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.

*Data acquisition software and connection cable sold separately— part number 37804-13.
Light sensor is sensitive to humidity; suggest storage is in the included case with a humidity sponge like 07193-11.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat