Trajan SGE eVol® XR MEPS Replacement Syringe, 100 µL, 25G, Bevel Tip; 3/Pk
Representative image only.
Trajan Mfr # 2910329 – Item # EW-17940-16
Replacements and enhancements to make the most of your digital syringe system
  • Compatible with Trajan SGE eVol® XR digital syringes
PRODUCT OPTIONS
$409.00USD / Pkg of 3
SPECS & DESCRIPTION
Specifications & Description
  • Syringe TypeReplacement syringe
  • Syringe Capacity (µL)100
  • Needle Tip StyleBevel
  • Needle IncludedYes
  • Needle Gauge25
  • Needle Length (in)2
  • SterileNo
  • Compatible WithSGE eVol® XR digital syringes
  • PowerNone
  • DescriptionMEPS Replacement Syringe, 100 µL, 25G, Bevel Tip; 3/Pk
Key Features
  • Compatible with Trajan SGE eVol® XR digital syringes
More About this Item
Use these accessories to optimize the performance of your eVol XR digital syringes. Digital syringes sold separately.
XCHANGE™ analytical syringes can be easily and quickly changed allowing them to be dedicated to individual liquids, and methods, to prevent possible cross-contamination of reagents.
Available for sale in the Middle East, Europe, and United States only.