Trajan SGE eVoL XR Electronic Syringe Starter Kit
Representative image only.
Trajan Mfr # 2910200 – Item # EW-17940-22
World's first digital analytic syringes combine precision and speed
  • Program and store up to 98 steps of workflow
  • Password protection options for improved security
  • Save calibrations for each syringe, enabling standards compliance
  • Suitable for direct injection onto a chromatography column with consistent flow rate
  • Be safer when working with volatile samples — aliquots are dispensed into sealed vials
SPECS & DESCRIPTION
Specifications & Description
  • Syringe TypeElectronic
  • Syringe Capacity (mL)1
  • Syringe Capacity (µL)100
  • Needle Tip StyleBevel
  • Needle Length (in)1.969
  • DescriptionEVOL XR Electronic Syringe Starter Kit
Key Features
  • Program and store up to 98 steps of workflow
  • Password protection options for improved security
  • Save calibrations for each syringe, enabling standards compliance
  • Suitable for direct injection onto a chromatography column with consistent flow rate
  • Be safer when working with volatile samples — aliquots are dispensed into sealed vials
More About this Item
eVol XR syringes combine a digitally controlled electronic drive and XCHANGE® enabled analytics for improved accuracy and reproducibility. The eVol XR system allows quick and simple syringe changes, so syringes can be dedicated to types of samples or methods, helping prevent reagent cross-contamination. And calibration factors can be easily programmed and stored for each syringe, enabling compliance with global lab standards. Syringe programming can be password protected for security.
eVol XR Starter Kit includes 5 µL, 100 µL, and 1 mL syringes with 5 cm needles.
Available for sale in the Middle East, Europe, and United States only.