Trajan SGE EVOL XR Replacement Syringe, 1 mL, 23G, Bevel Tip; 3/pk
Representative image only.
Trajan Mfr # 2910335 – Item # EW-17940-17
Digitally controlled electronic drive and an analytical syringe—all in one!
  • Easily and quickly change syringe with specialized connection system
  • Store laboratory workflow—up to 98 steps!
  • Use for both GC and LC injections
  • Volume: 1 mL Needle: 23G, Bevel Tip Pack size: 3
SPECS & DESCRIPTION
Specifications & Description
  • Syringe Capacity (µL)1000
  • Needle IncludedYes
  • Needle Tip StyleBevel
  • Needle Gauge23
  • SterileNo
  • DescriptionXR Replacement Syringe, 1 mL, 23G, Bevel Tip; 3/pk
Key Features
  • Easily and quickly change syringe with specialized connection system
  • Store laboratory workflow—up to 98 steps!
  • Use for both GC and LC injections
  • Volume: 1 mL Needle: 23G, Bevel Tip Pack size: 3
More About this Item
Create a more efficient workflow using these programmable digital syringes. Easily change XCHANGE® syringes allowing them to be dedicated to individual liquids or methods, preventing possible cross-contamination. Calibration and password protection ensure laboratory compliance. Use to dispense both aqueous and nonaqueous liquids. Stainless steel needle lets you inject directly through septa and chromatography columns with a consistent flow rate. Touch-wheel interface and full-color screen makes it easy to program and use.
Create a more efficient workflow using these programmable digital syringes. Easily exchange syringes allowing them to be dedicated to individual liquids or methods, preventing possible cross-contamination. Calibration feature and password protection feature ensure laboratory compliance. Use to dispense both aqueous and nonaqueous liquids. Stainless steel needle lets you inject directly through septa and chromatography columns with a consistent flow rate. Touch-wheel interface and full-color screen makes it easy to program and use.
Available for sale in the Middle East, Europe, and United States only.